The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_015258.1 | NP_056073.1 | 14 - 15 | 1497 | 91 |
XM_006717018.2 | XP_006717081.1 | 14 - 15 | 1414 | 91 | |
XM_006717019.1 | XP_006717082.1 | 13 - 14 | 1345 | 91 | |
XM_017014552.1 | XP_016870041.1 | 13 - 14 | 1345 | 91 | |
GenBank mRNA | AB014574.1 | - | 14 - 15 | 1424 | 91 |
AK301208.1 | - | 1 - 2 | 251 | 91 | |
AK303074.1 | - | 13 - 14 | 1345 | 91 | |
AK307498.1 | - | 13 - 14 | 1326 | 91 | |
AK307507.1 | - | 18 - 19 | 1931 | 91 | |
DA238568.1 | - | 5 - 6 | 477 | 91 |