The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_015086.1 | NP_055901.2 | 1 - 2 | 229 | 113 |
XM_011538055.2 | XP_011536357.1 | 2 - 3 | 834 | 113 | |
GenBank mRNA | AB018292.1 | - | 1 - 2 | 113 | 113 |
BC136398.1 | - | 1 - 2 | 139 | 113 | |
BC136399.1 | - | 1 - 2 | 139 | 113 |