The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001130965.2 | NP_001124437.1 | 6 - 7 | 777 | 95 |
GenBank mRNA | AB018353.1 | - | 7 - 8 | 875 | 95 |
AB648918.1 | - | 8 - 9 | 1066 | 95 | |
AK025737.1 | - | 3 - 4 | 302 | 95 | |
BC142707.1 | - | 6 - 7 | 787 | 95 | |
BC152418.1 | - | 6 - 7 | 875 | 95 | |
BX649069.1 | - | 4 - 5 | 532 | 95 |