Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
PLEKHM2
Entrez Gene ID
23207
Gene Name
pleckstrin homology and RUN domain containing M2
Gene Aliases
SKIP
Chromosome Location
Chr.1: 15681506 - 15734769 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_015164.2 NP_055979.2 1 - 2 287 72
XM_005245790.3 XP_005245847.1 1 - 2 482 72
GenBank mRNA AB020649.1 - 1 - 2 63 72
BC040441.1 - 1 - 2 290 72

View Full Product Details