Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
MYO16
Entrez Gene ID
23026
Gene Name
myosin XVI
Gene Aliases
MYAP3, MYR8, Myo16b, NYAP3, PPP1R107
Chromosome Location
Chr.13: 108495729 - 109208007 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001198950.1 NP_001185879.1 29 - 30 4150 73
NM_015011.1 NP_055826.1 29 - 30 3588 73
XM_011521062.1 XP_011519364.1 29 - 30 3696 73
GenBank mRNA AB020672.2 - 29 - 30 3588 73
AB290159.1 - 29 - 30 4151 73
AK302933.1 - 17 - 18 2291 73
BC146791.1 - 29 - 30 3588 73

View Full Product Details