The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001198950.1 | NP_001185879.1 | 32 - 33 | 5784 | 72 |
NM_015011.1 | NP_055826.1 | 32 - 33 | 5222 | 72 | |
GenBank mRNA | AB020672.2 | - | 32 - 33 | 5222 | 72 |
AB290159.1 | - | 32 - 33 | 5785 | 72 | |
BC146791.1 | - | 32 - 33 | 5222 | 72 |