Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
RALGAPA1
Entrez Gene ID
253959
Gene Name
Ral GTPase activating protein catalytic alpha subunit 1
Gene Aliases
GARNL1, GRIPE, RalGAPalpha1, TULIP1, p240
Chromosome Location
Chr.14: 35538352 - 35809304 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001283043.1 NP_001269972.1 9 - 10 1403 111
NM_001283044.1 NP_001269973.1 9 - 10 1403 111
NM_014990.1 NP_055805.1 9 - 10 1403 111
NM_194301.2 NP_919277.2 9 - 10 1403 111
XM_005267491.3 XP_005267548.1 9 - 10 1481 111
XM_005267492.3 XP_005267549.1 9 - 10 1481 111
XM_005267493.3 XP_005267550.1 9 - 10 1481 111
XM_006720098.2 XP_006720161.1 9 - 10 1481 111
XM_006720099.2 XP_006720162.1 9 - 10 1481 111
XM_006720100.3 XP_006720163.1 9 - 10 1481 111
XM_006720101.2 XP_006720164.1 9 - 10 1481 111
XM_006720102.3 XP_006720165.1 9 - 10 1187 111
XM_006720103.2 XP_006720166.1 9 - 10 1481 111
XM_006720104.3 XP_006720167.1 9 - 10 1481 111
XM_011536615.2 XP_011534917.1 9 - 10 1481 111
XM_011536616.2 XP_011534918.1 9 - 10 1481 111
XM_017021141.1 XP_016876630.1 9 - 10 1481 111
XM_017021142.1 XP_016876631.1 9 - 10 1481 111
XM_017021143.1 XP_016876632.1 9 - 10 1481 111
XM_017021144.1 XP_016876633.1 9 - 10 1481 111
XM_017021145.1 XP_016876634.1 9 - 10 1481 111
GenBank mRNA AB020691.1 - 4 - 5 531 111
AB511280.1 - 9 - 10 1012 111
AY596970.1 - 9 - 10 1403 111
AY596971.1 - 9 - 10 1403 111
BC016305.1 - 4 - 5 547 111
BC150596.1 - 9 - 10 1111 111

View Full Product Details