The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_015981.3 | NP_057065.2 | 11 - 12 | 1117 | 69 |
NM_171825.2 | NP_741960.1 | 11 - 12 | 1117 | 69 | |
XM_017009898.1 | XP_016865387.1 | 11 - 12 | 1117 | 69 | |
GenBank mRNA | AB023185.1 | - | 11 - 12 | 1050 | 69 |
AF091486.1 | - | 11 - 12 | 1006 | 69 | |
AF145710.1 | - | 11 - 12 | 908 | 69 | |
AF145711.1 | - | 11 - 12 | 908 | 69 | |
AK289776.1 | - | 11 - 12 | 1117 | 69 | |
AK308853.1 | - | 10 - 11 | 990 | 69 | |
BC040457.1 | - | 11 - 12 | 1032 | 69 |