The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001297716.1 | NP_001284645.1 | 4 - 5 | 1116 | 100 |
NM_014979.3 | NP_055794.3 | 4 - 5 | 1116 | 100 | |
XM_011543281.2 | XP_011541583.1 | 4 - 5 | 1483 | 100 | |
XM_011543282.2 | XP_011541584.2 | 4 - 5 | 2043 | 100 | |
GenBank mRNA | AB028977.1 | - | 2 - 3 | 177 | 100 |
AK094917.1 | - | 4 - 5 | 1117 | 100 | |
BC100824.1 | - | 4 - 5 | 1052 | 100 | |
BC100825.1 | - | 4 - 5 | 1050 | 100 | |
BC100826.1 | - | 4 - 5 | 1050 | 100 | |
BC100827.1 | - | 4 - 5 | 1050 | 100 |