The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_015168.1 | NP_055983.1 | 12 - 13 | 1837 | 112 |
XM_005258676.3 | XP_005258733.1 | 11 - 12 | 3112 | 112 | |
XM_005258677.4 | XP_005258734.1 | 12 - 13 | 1963 | 112 | |
XM_005258678.2 | XP_005258735.1 | 11 - 12 | 1720 | 112 | |
XM_006723113.3 | XP_006723176.1 | 12 - 13 | 3527 | 112 | |
XM_011526668.2 | XP_011524970.1 | 12 - 13 | 1739 | 112 | |
XM_011526669.2 | XP_011524971.1 | 11 - 12 | 1867 | 112 | |
XM_011526670.2 | XP_011524972.1 | 11 - 12 | 2463 | 112 | |
XM_017026529.1 | XP_016882018.1 | 11 - 12 | 2919 | 112 | |
XM_017026530.1 | XP_016882019.1 | 12 - 13 | 3527 | 112 | |
XM_017026531.1 | XP_016882020.1 | 10 - 11 | 1618 | 112 | |
GenBank mRNA | AB028987.2 | - | 12 - 13 | 1837 | 112 |