The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001323614.1 | NP_001310543.1 | 2 - 3 | 384 | 113 |
NM_001323615.1 | NP_001310544.1 | 2 - 3 | 384 | 113 | |
NM_001323616.1 | NP_001310545.1 | 3 - 4 | 516 | 113 | |
NM_001323617.1 | NP_001310546.1 | 3 - 4 | 351 | 113 | |
NM_015327.2 | NP_056142.2 | 2 - 3 | 323 | 113 | |
XM_017000843.1 | XP_016856332.1 | 2 - 3 | 330 | 113 | |
GenBank mRNA | AB029012.2 | - | 2 - 3 | 325 | 113 |
AB085691.1 | - | 2 - 3 | 251 | 113 | |
AK092183.1 | - | 2 - 3 | 299 | 113 | |
AY168922.1 | - | 2 - 3 | 177 | 113 | |
AY336728.1 | - | 2 - 3 | 174 | 113 | |
BC038296.1 | - | 2 - 3 | 324 | 113 | |
CT000143.1 | - | 2 - 3 | 384 | 113 |