Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
MYT1L
Entrez Gene ID
23040
Gene Name
myelin transcription factor 1 like
Gene Aliases
MRD39, NZF1, ZC2H2C2, ZC2HC4B
Chromosome Location
Chr.2: 1789113 - 2331388 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001303052.1 NP_001289981.1 19 - 20 3640 92
NM_015025.3 NP_055840.2 19 - 20 3634 92
XM_011510318.1 XP_011508620.1 20 - 21 3765 92
XM_011510319.1 XP_011508621.1 20 - 21 3875 92
XM_011510320.2 XP_011508622.1 20 - 21 3806 92
XM_011510321.2 XP_011508623.1 20 - 21 3756 92
XM_011510322.2 XP_011508624.1 20 - 21 3569 92
XM_011510323.1 XP_011508625.1 19 - 20 3488 92
XM_011510324.1 XP_011508626.1 20 - 21 3765 92
XM_011510325.1 XP_011508627.1 20 - 21 3759 92
XM_011510326.1 XP_011508628.1 19 - 20 3702 92
XM_011510327.1 XP_011508629.1 20 - 21 3765 92
XM_011510328.2 XP_011508630.1 20 - 21 3765 92
XM_017003604.1 XP_016859093.1 21 - 22 3854 92
XM_017003605.1 XP_016859094.1 20 - 21 3433 92
XM_017003606.1 XP_016859095.1 20 - 21 3869 92
XM_017003607.1 XP_016859096.1 20 - 21 3800 92
XM_017003608.1 XP_016859097.1 20 - 21 3765 92
XM_017003609.1 XP_016859098.1 19 - 20 3687 92
XM_017003610.1 XP_016859099.1 20 - 21 3786 92
XM_017003611.1 XP_016859100.1 19 - 20 3696 92
XM_017003612.1 XP_016859101.1 19 - 20 3806 92
XM_017003613.1 XP_016859102.1 19 - 20 3806 92
XM_017003614.1 XP_016859103.1 19 - 20 3645 92
XM_017003615.1 XP_016859104.1 19 - 20 3702 92
XM_017003616.1 XP_016859105.1 19 - 20 3702 92
XM_017003617.1 XP_016859106.1 19 - 20 3696 92
XM_017003618.1 XP_016859107.1 19 - 20 3696 92
XM_017003619.1 XP_016859108.1 19 - 20 3806 92
XM_017003620.1 XP_016859109.1 13 - 14 2755 92
GenBank mRNA AB029029.2 - 20 - 21 3419 92
BC031690.1 - 11 - 12 2383 92
BC137272.1 - 19 - 20 3437 92
BC137273.1 - 19 - 20 3437 92
BC150281.1 - 20 - 21 3419 92

View Full Product Details