Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
DOPEY1
Entrez Gene ID
23033
Gene Name
dopey family member 1
Gene Aliases
DOP1, KIAA1117, dJ202D23.2
Chromosome Location
Chr.6: 83067650 - 83171351 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001199942.1 NP_001186871.1 35 - 36 6971 81
NM_015018.3 NP_055833.2 35 - 36 6998 81
XM_011535619.2 XP_011533921.1 34 - 35 6963 81
XM_017010559.1 XP_016866048.1 35 - 36 6993 81
XM_017010560.1 XP_016866049.1 34 - 35 6921 81
XM_017010561.1 XP_016866050.1 35 - 36 6945 81
XM_017010562.1 XP_016866051.1 34 - 35 6894 81
XM_017010563.1 XP_016866052.1 35 - 36 6972 81
XM_017010564.1 XP_016866053.1 35 - 36 6972 81
XM_017010565.1 XP_016866054.1 35 - 36 6966 81
XM_017010566.1 XP_016866055.1 35 - 36 6966 81
XM_017010567.1 XP_016866056.1 34 - 35 6894 81
XM_017010568.1 XP_016866057.1 34 - 35 6963 81
XM_017010569.1 XP_016866058.1 33 - 34 6880 81
XM_017010570.1 XP_016866059.1 34 - 35 6894 81
XM_017010571.1 XP_016866060.1 34 - 35 6888 81
XM_017010572.1 XP_016866061.1 34 - 35 6843 81
XM_017010573.1 XP_016866062.1 35 - 36 6993 81
XM_017010574.1 XP_016866063.1 35 - 36 6993 81
GenBank mRNA AB029040.2 - 18 - 19 4019 81
AK027030.1 - 15 - 16 2765 81
AL050080.1 - 7 - 8 727 81
BC048342.1 - 9 - 10 1011 81
BC144544.1 - 35 - 36 6869 81
BC150619.1 - 35 - 36 6869 81
BX648667.1 - 35 - 36 6996 81

View Full Product Details