The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_020700.1 | NP_065751.1 | 7 - 8 | 1289 | 88 |
XM_011538578.2 | XP_011536880.1 | 7 - 8 | 1425 | 88 | |
XM_011538579.1 | XP_011536881.1 | 7 - 8 | 977 | 88 | |
XM_017019676.1 | XP_016875165.1 | 7 - 8 | 1539 | 88 | |
GenBank mRNA | AB032983.1 | - | 6 - 7 | 882 | 88 |
AB084258.1 | - | 7 - 8 | 1554 | 88 | |
AK023315.1 | - | 8 - 9 | 1351 | 88 | |
BC157843.1 | - | 7 - 8 | 1299 | 88 |