The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_020718.3 | NP_065769.3 | 15 - 16 | 2492 | 66 |
XM_017023496.1 | XP_016878985.1 | 14 - 15 | 2403 | 66 | |
XM_017023497.1 | XP_016878986.1 | 15 - 16 | 4018 | 66 | |
XM_017023498.1 | XP_016878987.1 | 14 - 15 | 1805 | 66 | |
XM_017023499.1 | XP_016878988.1 | 12 - 13 | 1609 | 66 | |
GenBank mRNA | AB033029.1 | - | 4 - 5 | 617 | 66 |
AJ586135.2 | - | 15 - 16 | 2492 | 66 | |
AK092790.1 | - | 1 - 2 | 508 | 66 | |
AK126752.1 | - | 15 - 16 | 2176 | 66 | |
BX648357.1 | - | 6 - 7 | 830 | 66 |