Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
PRR12
Entrez Gene ID
57479
Gene Name
proline rich 12
Gene Aliases
KIAA1205
Chromosome Location
Chr.19: 49591177 - 49626439 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_017027031.1 XP_016882520.1 6 - 7 4707 74
GenBank mRNA AB033031.1 - 5 - 6 2439 74
BC034003.1 - 3 - 4 805 74

View Full Product Details