The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001282917.1 | NP_001269846.1 | 23 - 24 | 3682 | 108 |
NM_001282918.1 | NP_001269847.1 | 23 - 24 | 3812 | 108 | |
NM_020336.3 | NP_065069.1 | 23 - 24 | 3824 | 108 | |
XM_005260462.2 | XP_005260519.1 | 24 - 25 | 3818 | 108 | |
XM_005260463.2 | XP_005260520.1 | 24 - 25 | 3818 | 108 | |
XM_005260464.2 | XP_005260521.1 | 24 - 25 | 3806 | 108 | |
XM_005260465.2 | XP_005260522.1 | 23 - 24 | 3782 | 108 | |
XM_017027966.1 | XP_016883455.1 | 24 - 25 | 3806 | 108 | |
XM_017027967.1 | XP_016883456.1 | 23 - 24 | 3770 | 108 | |
XM_017027968.1 | XP_016883457.1 | 23 - 24 | 3344 | 108 | |
XM_017027969.1 | XP_016883458.1 | 24 - 25 | 3835 | 108 | |
XM_017027970.1 | XP_016883459.1 | 8 - 9 | 1306 | 108 | |
GenBank mRNA | AB033045.3 | - | 23 - 24 | 3802 | 108 |
AK125260.1 | - | 11 - 12 | 1585 | 108 | |
AL834436.1 | - | 21 - 22 | 3046 | 108 | |
BC018668.1 | - | 2 - 3 | 317 | 108 | |
BC047482.1 | - | 23 - 24 | 3787 | 108 | |
CR936800.1 | - | 23 - 24 | 3588 | 108 |