Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
HEG1
Entrez Gene ID
57493
Gene Name
heart development protein with EGF like domains 1
Gene Aliases
HEG, MST112, MSTP112
Chromosome Location
Chr.3: 124965710 - 125055958 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_020733.1 NP_065784.1 7 - 8 3135 64
XM_005247666.1 XP_005247723.1 8 - 9 3430 64
GenBank mRNA AB033063.2 - 6 - 7 2730 64

View Full Product Details