Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
MKL2
Entrez Gene ID
57496
Gene Name
MKL1/myocardin like 2
Gene Aliases
MRTF-B, MRTFB, NPD001
Chromosome Location
Chr.16: 14071076 - 14266779 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001308142.1 NP_001295071.1 10 - 11 1256 98
NM_014048.4 NP_054767.3 10 - 11 1257 98
XM_005255452.3 XP_005255509.1 11 - 12 1411 98
XM_005255453.4 XP_005255510.1 10 - 11 1895 98
XM_005255455.3 XP_005255512.1 8 - 9 1241 98
XM_006720908.3 XP_006720971.1 11 - 12 2057 98
XM_006720909.3 XP_006720972.1 11 - 12 1386 98
XM_006720912.3 XP_006720975.1 8 - 9 3196 98
XM_006720913.2 XP_006720976.1 11 - 12 1411 98
XM_006720914.2 XP_006720977.1 11 - 12 1411 98
XM_011522568.2 XP_011520870.1 10 - 11 1226 98
XM_011522569.2 XP_011520871.1 8 - 9 1023 98
XM_017023501.1 XP_016878990.1 10 - 11 1222 98
XM_017023502.1 XP_016878991.1 10 - 11 1279 98
XM_017023503.1 XP_016878992.1 9 - 10 4424 98
XM_017023504.1 XP_016878993.1 8 - 9 1241 98
XM_017023505.1 XP_016878994.1 8 - 9 3196 98
GenBank mRNA AB033069.2 - 2 - 3 263 98
AK294765.1 - 10 - 11 1248 98
BC034934.1 - 9 - 10 1209 98
BC136260.1 - 8 - 9 1101 98
BC171750.1 - 8 - 9 1101 98

View Full Product Details