Assay Design
The assay probe spans an exon junction.
Gene Transcripts
- Gene Symbol
- GDA
- Entrez Gene ID
-
9615
- Gene Name
- guanine deaminase
- Gene Aliases
-
CYPIN, GUANASE, NEDASIN
- Chromosome Location
-
Chr.9: 72114595 - 72256045 on Build GRCh38
View Full Product Details