The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_017553.2 | NP_060023.1 | 28 - 29 | 3815 | 66 |
NR_104038.1 | 27 - 28 | 3687 | 66 | ||
XM_011521686.2 | XP_011519988.1 | 14 - 15 | 1626 | 66 | |
GenBank mRNA | AB033085.1 | - | 28 - 29 | 3711 | 66 |
AK026050.1 | - | 9 - 10 | 1165 | 66 | |
AK057582.1 | - | 17 - 18 | 2080 | 66 | |
AK290371.1 | - | 27 - 28 | 3553 | 66 | |
BC144681.1 | - | 27 - 28 | 3573 | 66 | |
BC146785.1 | - | 28 - 29 | 3723 | 66 |