Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
SLC39A10
Entrez Gene ID
57181
Gene Name
solute carrier family 39 member 10
Gene Aliases
LZT-Hs2
Chromosome Location
Chr.2: 195575949 - 195737702 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001127257.1 NP_001120729.1 7 - 8 2280 66
NM_020342.2 NP_065075.1 7 - 8 2231 66
XM_005246689.4 XP_005246746.2 7 - 8 2238 66
XM_011511504.2 XP_011509806.1 7 - 8 2145 66
XM_011511505.2 XP_011509807.1 8 - 9 2309 66
XM_011511506.2 XP_011509808.1 7 - 8 4968 66
XM_011511507.2 XP_011509809.1 7 - 8 2238 66
XM_017004522.1 XP_016860011.1 7 - 8 3270 66
GenBank mRNA AB033091.1 - 7 - 8 2144 66
AK074996.1 - 1 - 2 93 66
AK291241.1 - 7 - 8 2143 66
AK294771.1 - 8 - 9 1327 66
BC073909.1 - 3 - 4 656 66
BC101516.1 - 7 - 8 2140 66
BC112223.1 - 7 - 8 2152 66
CR749813.1 - 7 - 8 2131 66

View Full Product Details