The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001314030.1 | NP_001300959.1 | 3 - 4 | 701 | 85 |
NM_012284.2 | NP_036416.1 | 3 - 4 | 705 | 85 | |
XM_011538085.2 | XP_011536387.1 | 3 - 4 | 1321 | 85 | |
XM_011538086.2 | XP_011536388.1 | 3 - 4 | 1317 | 85 | |
XM_017019096.1 | XP_016874585.1 | 2 - 3 | 1087 | 85 | |
GenBank mRNA | AB022696.1 | - | 3 - 4 | 480 | 85 |
AB033108.1 | - | 3 - 4 | 705 | 85 | |
BC150602.1 | - | 3 - 4 | 586 | 85 |