The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001300906.1 | NP_001287835.1 | 4 - 5 | 1092 | 107 |
NM_015525.3 | NP_056340.2 | 4 - 5 | 1092 | 107 | |
XM_006715453.2 | XP_006715516.1 | 4 - 5 | 1632 | 107 | |
XM_006715454.2 | XP_006715517.1 | 4 - 5 | 1632 | 107 | |
GenBank mRNA | AB037838.2 | - | 3 - 4 | 816 | 107 |
BC092410.1 | - | 3 - 4 | 692 | 107 | |
BC113696.1 | - | 3 - 4 | 621 | 107 | |
BC113698.1 | - | 3 - 4 | 621 | 107 | |
BC143736.1 | - | 3 - 4 | 621 | 107 | |
BC143745.1 | - | 3 - 4 | 622 | 107 | |
BC143748.1 | - | 3 - 4 | 621 | 107 | |
DQ005633.1 | - | 4 - 5 | 1092 | 107 | |
DQ005634.1 | - | 4 - 5 | 1092 | 107 |