Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
ANO8
Entrez Gene ID
57719
Gene Name
anoctamin 8
Gene Aliases
KIAA1623, TMEM16H
Chromosome Location
Chr.19: 17323223 - 17334834 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_020959.2 NP_066010.1 2 - 3 375 56
XM_017027048.1 XP_016882537.1 2 - 3 380 56
GenBank mRNA AB046843.2 - 2 - 3 375 56

View Full Product Details