Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
ANKS3
Entrez Gene ID
124401
Gene Name
ankyrin repeat and sterile alpha motif domain containing 3
Gene Aliases
-
Chromosome Location
Chr.16: 4696510 - 4734377 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001242929.1 NP_001229858.1 5 - 6 776 93
NM_001308089.1 NP_001295018.1 7 - 8 966 93
NM_001324129.1 NP_001311058.1 4 - 5 604 93
NM_133450.3 NP_597707.1 8 - 9 1168 93
NR_040252.1 7 - 8 1171 93
XM_005255119.3 XP_005255176.1 6 - 7 868 93
XM_005255121.3 XP_005255178.1 4 - 5 566 93
XM_011522372.1 XP_011520674.1 7 - 8 1098 93
XM_011522373.1 XP_011520675.1 7 - 8 980 93
XM_011522374.1 XP_011520676.1 6 - 7 714 93
XM_011522378.1 XP_011520680.1 7 - 8 836 93
XM_017022935.1 XP_016878424.1 5 - 6 776 93
XM_017022936.1 XP_016878425.1 7 - 8 1089 93
GenBank mRNA AB075857.1 - 5 - 6 1604 93
AK057329.1 - 5 - 6 769 93
AK123371.1 - 4 - 5 565 93
AK123496.1 - 8 - 9 1169 93
AK301683.1 - 6 - 7 796 93
BC050387.1 - 7 - 8 807 93
BX333650.2 - 5 - 6 621 93

View Full Product Details