The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_133452.2 | NP_597709.2 | 10 - 11 | 1978 | 133 |
XM_011527671.1 | XP_011525973.1 | 11 - 12 | 1897 | 133 | |
GenBank mRNA | AB075858.1 | - | 7 - 8 | 3081 | 133 |
BC037565.1 | - | 8 - 9 | 1315 | 133 | |
CR746909.1 | - | 2 - 3 | 282 | 133 |