The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_181844.3 | NP_862827.1 | 5 - 6 | 957 | 71 |
GenBank mRNA | AB076580.1 | - | 5 - 6 | 937 | 71 |
AK122893.1 | - | 5 - 6 | 953 | 71 | |
AK292878.1 | - | 5 - 6 | 940 | 71 | |
AK293102.1 | - | 5 - 6 | 956 | 71 |