The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_003502.3 | NP_003493.1 | 1 - 2 | 305 | 75 |
NM_181050.2 | NP_851393.1 | 1 - 2 | 305 | 75 | |
NR_134879.1 | 1 - 2 | 305 | 75 | ||
XM_011522682.2 | XP_011520984.1 | 1 - 2 | 316 | 75 | |
XM_011522683.2 | XP_011520985.1 | 1 - 2 | 315 | 75 | |
XM_017023743.1 | XP_016879232.1 | 1 - 2 | 316 | 75 | |
XM_017023744.1 | XP_016879233.1 | 1 - 2 | 315 | 75 | |
XM_017023747.1 | XP_016879236.1 | 1 - 2 | 318 | 75 | |
XM_017023748.1 | XP_016879237.1 | 1 - 2 | 318 | 75 | |
GenBank mRNA | AF009674.1 | - | 1 - 2 | 28 | 75 |
BC044648.1 | - | 1 - 2 | 77 | 75 | |
KF615850.1 | - | 1 - 2 | 305 | 75 | |
KF615851.1 | - | 1 - 2 | 305 | 75 | |
KF615852.1 | - | 1 - 2 | 305 | 75 |