The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_004685.3 | NP_004676.3 | 7 - 8 | 1182 | 93 |
XM_011535306.1 | XP_011533608.1 | 8 - 9 | 1242 | 93 | |
XM_011535307.1 | XP_011533609.1 | 8 - 9 | 1242 | 93 | |
XM_017020845.1 | XP_016876334.1 | 7 - 8 | 1128 | 93 | |
XM_017020846.1 | XP_016876335.1 | 4 - 5 | 447 | 93 | |
GenBank mRNA | AF072928.1 | - | 3 - 4 | 396 | 93 |
AF406619.1 | - | 7 - 8 | 863 | 93 | |
AK314587.1 | - | 7 - 8 | 1047 | 93 | |
AL832017.2 | - | 7 - 8 | 1105 | 93 | |
BC040012.1 | - | 7 - 8 | 1054 | 93 |