Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
SEPN1
Entrez Gene ID
57190
Gene Name
selenoprotein N, 1
Gene Aliases
CFTD, MDRS1, RSMD1, RSS, SELN
Chromosome Location
Chr.1: 25800176 - 25818222 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_020451.2 NP_065184.2 8 - 9 1148 78
NM_206926.1 NP_996809.1 7 - 8 1046 78
GenBank mRNA AF166125.1 - 6 - 7 737 78
AJ306399.1 - 8 - 9 1148 78
AK308457.1 - 6 - 7 766 78
BC021028.2 - 3 - 4 351 78
BQ217758.1 - 5 - 6 604 79

View Full Product Details