Assay Design
The assay probe spans an exon junction.
Gene Transcripts
- Gene Symbol
- BAGE2
- Entrez Gene ID
-
85319
- Gene Name
- B melanoma antigen family member 2
- Gene Aliases
-
CT2.2
- Chromosome Location
-
Chr.21: 10413532 - 10491615 on Build GRCh38
- Gene Symbol
- BAGE3
- Entrez Gene ID
-
85318
- Gene Name
- B melanoma antigen family member 3
- Gene Aliases
-
CT2.3
- Chromosome Location
-
-
- Gene Symbol
- BAGE4
- Entrez Gene ID
-
85317
- Gene Name
- B melanoma antigen family member 4
- Gene Aliases
-
CT2.4, MLL3P
- Chromosome Location
-
-
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_181704.1 |
NP_859055.1 |
4 - 5 |
725 |
79 |
GenBank mRNA |
AF339515.1 |
- |
4 - 5 |
725 |
79 |
- Gene Symbol
- BAGE5
- Entrez Gene ID
-
85316
- Gene Name
- B melanoma antigen family member 5
- Gene Aliases
-
CT2.5
- Chromosome Location
-
-
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_182484.1 |
NP_872290.1 |
4 - 5 |
725 |
79 |
GenBank mRNA |
AF339516.1 |
- |
4 - 5 |
725 |
79 |
- Gene Symbol
- KMT2C
- Entrez Gene ID
-
58508
- Gene Name
- lysine methyltransferase 2C
- Gene Aliases
-
HALR, MLL3
- Chromosome Location
-
Chr.7: 152134925 - 152436642 on Build GRCh38
- Gene Symbol
- LOC105378260
- Entrez Gene ID
-
105378260
- Gene Name
- histone-lysine N-methyltransferase 2C
- Gene Aliases
-
-
- Chromosome Location
-
Chr.21: 10462898 - 10514209 on Build GRCh38
View Full Product Details