The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001198999.1 | NP_001185928.1 | 13 - 14 | 1414 | 141 |
NM_020858.1 | NP_065909.1 | 10 - 11 | 1402 | 141 | |
NM_024966.2 | NP_079242.2 | 10 - 11 | 1402 | 141 | |
NM_153616.1 | NP_705869.1 | 10 - 11 | 1402 | 141 | |
NM_153617.1 | NP_705870.1 | 10 - 11 | 1402 | 141 | |
NM_153618.1 | NP_705871.1 | 10 - 11 | 1402 | 141 | |
NM_153619.1 | NP_705872.1 | 10 - 11 | 1402 | 141 | |
XM_005254685.3 | XP_005254742.1 | 10 - 11 | 1364 | 141 | |
XM_005254686.2 | XP_005254743.1 | 10 - 11 | 1308 | 141 | |
XM_005254687.2 | XP_005254744.1 | 10 - 11 | 1308 | 141 | |
XM_005254689.3 | XP_005254746.1 | 10 - 11 | 1364 | 141 | |
XM_011522075.2 | XP_011520377.1 | 13 - 14 | 1519 | 141 | |
XM_011522076.2 | XP_011520378.1 | 10 - 11 | 1308 | 141 | |
XM_011522077.2 | XP_011520379.1 | 10 - 11 | 1308 | 141 | |
XM_011522078.2 | XP_011520380.1 | 10 - 11 | 1308 | 141 | |
XM_011522079.2 | XP_011520381.1 | 10 - 11 | 1308 | 141 | |
XM_011522080.2 | XP_011520382.1 | 10 - 11 | 1308 | 141 | |
XM_011522081.2 | XP_011520383.1 | 10 - 11 | 1308 | 141 | |
XM_017022617.1 | XP_016878106.1 | 13 - 14 | 1332 | 141 | |
XM_017022618.1 | XP_016878107.1 | 10 - 11 | 1364 | 141 | |
XM_017022619.1 | XP_016878108.1 | 10 - 11 | 1364 | 141 | |
XM_017022620.1 | XP_016878109.1 | 13 - 14 | 1519 | 141 | |
XM_017022621.1 | XP_016878110.1 | 13 - 14 | 1519 | 141 | |
GenBank mRNA | AB040912.2 | - | 13 - 14 | 1414 | 141 |
AF389426.1 | - | 10 - 11 | 1402 | 141 | |
AF389427.1 | - | 10 - 11 | 1402 | 141 | |
AF389428.1 | - | 10 - 11 | 1402 | 141 | |
AF389429.1 | - | 10 - 11 | 1402 | 141 | |
AF389430.1 | - | 10 - 11 | 1402 | 141 | |
AK021660.1 | - | 7 - 8 | 700 | 141 | |
AK290032.1 | - | 10 - 11 | 1323 | 141 | |
BC150253.1 | - | 13 - 14 | 1414 | 141 |