The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001290258.1 | NP_001277187.1 | 11 - 12 | 1884 | 71 |
NM_080928.4 | NP_563616.3 | 9 - 10 | 1669 | 71 | |
XM_005250146.4 | XP_005250203.1 | 11 - 12 | 2004 | 71 | |
XM_011515816.2 | XP_011514118.1 | 9 - 10 | 1665 | 71 | |
XM_011515817.2 | XP_011514119.1 | 12 - 13 | 2065 | 71 | |
XM_011515818.2 | XP_011514120.1 | 12 - 13 | 2102 | 71 | |
XM_011515819.2 | XP_011514121.1 | 12 - 13 | 2047 | 71 | |
XM_011515820.2 | XP_011514122.1 | 10 - 11 | 1879 | 71 | |
XM_011515821.2 | XP_011514123.1 | 8 - 9 | 1506 | 71 | |
XM_017011756.1 | XP_016867245.1 | 12 - 13 | 2950 | 71 | |
XM_017011757.1 | XP_016867246.1 | 12 - 13 | 2180 | 71 | |
XM_017011758.1 | XP_016867247.1 | 5 - 6 | 1228 | 71 | |
GenBank mRNA | AF403033.1 | - | 4 - 5 | 1131 | 71 |
AF428257.1 | - | 10 - 11 | 1425 | 71 | |
AK125360.1 | - | 11 - 12 | 1884 | 71 | |
CR749595.1 | - | 6 - 7 | 1411 | 71 |