Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
FOXN4
Entrez Gene ID
121643
Gene Name
forkhead box N4
Gene Aliases
-
Chromosome Location
Chr.12: 109277978 - 109309406 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_213596.2 NP_998761.2 6 - 7 704 70
XM_011537922.2 XP_011536224.1 6 - 7 893 70
XM_011537923.2 XP_011536225.1 6 - 7 893 70
XM_017018818.1 XP_016874307.1 4 - 5 1679 70
XM_017018819.1 XP_016874308.1 6 - 7 893 70
GenBank mRNA AF425596.1 - 4 - 5 479 70
BC146825.1 - 6 - 7 705 70

View Full Product Details