Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
SYNE2
Entrez Gene ID
23224
Gene Name
spectrin repeat containing nuclear envelope protein 2
Gene Aliases
EDMD5, NUA, NUANCE, Nesp2, Nesprin-2, SYNE-2, TROPH
Chromosome Location
Chr.14: 63761904 - 64226449 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_015180.4 NP_055995.4 34 - 35 5272 139
NM_182914.2 NP_878918.2 34 - 35 5272 139
XM_005267454.1 XP_005267511.1 34 - 35 5272 139
XM_005267456.1 XP_005267513.1 34 - 35 5272 139
XM_005267457.1 XP_005267514.1 34 - 35 5272 139
XM_005267458.1 XP_005267515.1 34 - 35 5272 139
XM_005267459.1 XP_005267516.1 34 - 35 5272 139
XM_011536574.1 XP_011534876.1 34 - 35 5272 139
XM_011536575.2 XP_011534877.1 34 - 35 5249 139
XM_011536576.2 XP_011534878.1 35 - 36 5429 139
XM_011536577.2 XP_011534879.1 34 - 35 5196 139
XM_011536578.1 XP_011534880.1 34 - 35 5272 139
XM_011536579.1 XP_011534881.1 34 - 35 5272 139
XM_011536580.1 XP_011534882.1 34 - 35 5272 139
XM_011536581.1 XP_011534883.1 34 - 35 5272 139
XM_011536582.1 XP_011534884.1 34 - 35 5272 139
XM_011536584.2 XP_011534886.1 34 - 35 5272 139
XM_017021101.1 XP_016876590.1 34 - 35 5274 139
XM_017021102.1 XP_016876591.1 33 - 34 5317 139
GenBank mRNA AF357236.1 - 33 - 34 5042 139
AF435011.1 - 34 - 35 5254 139
AF495911.1 - 34 - 35 5239 139
AK125715.1 - 5 - 6 934 139
BX648234.1 - 5 - 6 952 139

View Full Product Details