The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_017539.2 | NP_060009.1 | 2 - 3 | 217 | 69 |
XM_006721062.1 | XP_006721125.1 | 3 - 4 | 406 | 69 | |
XM_011545882.2 | XP_011544184.1 | 4 - 5 | 846 | 69 | |
XM_011545883.1 | XP_011544185.1 | 4 - 5 | 614 | 69 | |
XM_011545885.2 | XP_011544187.1 | 4 - 5 | 846 | 69 | |
XM_017023426.1 | XP_016878915.1 | 4 - 5 | 846 | 69 | |
XM_017023427.1 | XP_016878916.1 | 4 - 5 | 846 | 69 | |
XM_017023428.1 | XP_016878917.1 | 4 - 5 | 846 | 69 | |
XM_017023429.1 | XP_016878918.1 | 4 - 5 | 847 | 69 | |
XM_017023431.1 | XP_016878920.1 | 4 - 5 | 847 | 69 | |
XM_017023432.1 | XP_016878921.1 | 2 - 3 | 313 | 69 | |
GenBank mRNA | AF494040.1 | - | 2 - 3 | 217 | 69 |
AK056509.2 | - | 3 - 4 | 406 | 69 | |
AK303958.1 | - | 3 - 4 | 415 | 69 | |
AK310973.1 | - | 3 - 4 | 403 | 69 |