The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_017539.2 | NP_060009.1 | 28 - 29 | 4036 | 91 |
XM_006721062.1 | XP_006721125.1 | 28 - 29 | 4174 | 91 | |
XM_011545882.2 | XP_011544184.1 | 29 - 30 | 4614 | 91 | |
XM_011545883.1 | XP_011544185.1 | 29 - 30 | 4382 | 91 | |
XM_011545885.2 | XP_011544187.1 | 29 - 30 | 4614 | 91 | |
XM_011545886.2 | XP_011544188.1 | 21 - 22 | 2989 | 91 | |
XM_011545888.2 | XP_011544190.1 | 11 - 12 | 1433 | 91 | |
XM_011545889.2 | XP_011544191.1 | 11 - 12 | 1397 | 91 | |
XM_017023426.1 | XP_016878915.1 | 29 - 30 | 4614 | 91 | |
XM_017023427.1 | XP_016878916.1 | 27 - 28 | 4371 | 91 | |
XM_017023428.1 | XP_016878917.1 | 29 - 30 | 4614 | 91 | |
XM_017023429.1 | XP_016878918.1 | 29 - 30 | 4615 | 91 | |
XM_017023431.1 | XP_016878920.1 | 29 - 30 | 4615 | 91 | |
XM_017023432.1 | XP_016878921.1 | 28 - 29 | 4174 | 91 | |
GenBank mRNA | AF494040.1 | - | 28 - 29 | 4036 | 91 |
AK125907.1 | - | 5 - 6 | 753 | 91 |