The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_145755.2 | NP_665698.2 | 7 - 8 | 1006 | 149 |
GenBank mRNA | AJ487015.1 | - | 7 - 8 | 968 | 149 |