Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
WNK2
Entrez Gene ID
65268
Gene Name
WNK lysine deficient protein kinase 2
Gene Aliases
NY-CO-43, P/OKcl.13, PRKWNK2, SDCCAG43
Chromosome Location
Chr.9: 93184156 - 93327581 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001282394.1 NP_001269323.1 24 - 25 6033 84
NM_006648.3 NP_006639.3 23 - 24 5922 84
XM_005252137.3 XP_005252194.1 24 - 25 6154 84
XM_005252140.2 XP_005252197.1 24 - 25 6154 84
XM_005252141.2 XP_005252198.1 24 - 25 6154 84
XM_005252144.2 XP_005252201.1 24 - 25 6154 84
XM_011518926.2 XP_011517228.1 24 - 25 6154 84
XM_011518927.2 XP_011517229.1 24 - 25 6151 84
XM_011518929.2 XP_011517231.1 24 - 25 6154 84
XM_011518930.2 XP_011517232.1 24 - 25 6019 84
XM_011518931.2 XP_011517233.1 24 - 25 6154 84
XM_011518932.2 XP_011517234.1 24 - 25 6154 84
XM_011518937.2 XP_011517239.1 23 - 24 5462 84
XM_011518939.2 XP_011517241.1 23 - 24 5507 84
XM_011518940.2 XP_011517242.1 23 - 24 5509 84
XM_017015045.1 XP_016870534.1 24 - 25 6151 84
XM_017015046.1 XP_016870535.1 24 - 25 6016 84
XM_017015049.1 XP_016870538.1 24 - 25 6154 84
XM_017015050.1 XP_016870539.1 24 - 25 6151 84
XM_017015051.1 XP_016870540.1 24 - 25 6019 84
GenBank mRNA AB051547.2 - 23 - 24 5909 84
AJ242724.4 - 24 - 25 6033 84
AK000694.1 - 9 - 10 2312 84
BC037965.1 - 5 - 6 1221 84

View Full Product Details