Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
NEMF
Entrez Gene ID
9147
Gene Name
nuclear export mediator factor
Gene Aliases
NY-CO-1, SDCCAG1
Chromosome Location
Chr.14: 49782083 - 49853073 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_011537317.2 XP_011535619.1 3 - 4 524 75
XM_011537318.2 XP_011535620.1 3 - 4 524 75
XM_017021760.1 XP_016877249.1 3 - 4 301 75
GenBank mRNA AK000913.1 - 3 - 4 255 75
AK295010.1 - 3 - 4 263 75
AK308537.1 - 3 - 4 260 75
BC056687.1 - 3 - 4 271 75
BM835709.1 - 3 - 4 300 75

View Full Product Details