Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.15: 42723220-42723723 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CDAN1
Entrez Gene ID
146059
Gene Name
codanin 1
Gene Aliases
CDA1, CDAI, CDAN1A, DLT, PRO1295
UniGene
Hs.599232
Gene Chromosome Location
Chr.15: 42723557-42737219 on Build GRCh38
Transcripts
NM_138477.2 XM_005254176.4 XM_011521270.2 XM_011521271.2 XM_011521274.2

SNP Detail

SNP ID Chromosome Location
rs1034847267 Chr.15: 42723503-42723503 on Build GRCh38
rs566104187 Chr.15: 42723361-42723361 on Build GRCh38
rs942800593 Chr.15: 42723393-42723393 on Build GRCh38
rs901390343 Chr.15: 42723405-42723405 on Build GRCh38
rs1030623435 Chr.15: 42723290-42723290 on Build GRCh38
rs925447749 Chr.15: 42723253-42723253 on Build GRCh38
rs931271817 Chr.15: 42723385-42723385 on Build GRCh38
rs1036317292 Chr.15: 42723364-42723368 on Build GRCh38
rs776456107 Chr.15: 42723427-42723427 on Build GRCh38
rs921265026 Chr.15: 42723634-42723634 on Build GRCh38
rs1012674382 Chr.15: 42723685-42723685 on Build GRCh38
rs961903811 Chr.15: 42723516-42723516 on Build GRCh38
rs1012624226 Chr.15: 42723473-42723473 on Build GRCh38
rs988226389 Chr.15: 42723553-42723553 on Build GRCh38
rs999757126 Chr.15: 42723262-42723262 on Build GRCh38
rs911254347 Chr.15: 42723618-42723618 on Build GRCh38
rs959674537 Chr.15: 42723436-42723436 on Build GRCh38
rs1041178708 Chr.15: 42723400-42723400 on Build GRCh38
rs556730759 Chr.15: 42723592-42723592 on Build GRCh38
rs999724655 Chr.15: 42723435-42723435 on Build GRCh38
rs889844076 Chr.15: 42723391-42723391 on Build GRCh38
rs768374518 Chr.15: 42723285-42723285 on Build GRCh38
rs766922997 Chr.15: 42723638-42723638 on Build GRCh38
rs989169817 Chr.15: 42723306-42723306 on Build GRCh38
rs1048392347 Chr.15: 42723390-42723390 on Build GRCh38
rs984498307 Chr.15: 42723576-42723576 on Build GRCh38
rs942785975 Chr.15: 42723665-42723665 on Build GRCh38
rs944032215 Chr.15: 42723610-42723610 on Build GRCh38
rs1004476592 Chr.15: 42723647-42723647 on Build GRCh38
rs138478605 Chr.15: 42723539-42723539 on Build GRCh38
rs918586101 Chr.15: 42723364-42723364 on Build GRCh38
rs369460870 Chr.15: 42723687-42723687 on Build GRCh38
rs560005758 Chr.15: 42723362-42723362 on Build GRCh38
rs1039584107 Chr.15: 42723331-42723331 on Build GRCh38
rs957978101 Chr.15: 42723296-42723296 on Build GRCh38
rs957113519 Chr.15: 42723540-42723540 on Build GRCh38
rs746543113 Chr.15: 42723244-42723244 on Build GRCh38
rs1052962601 Chr.15: 42723286-42723286 on Build GRCh38
rs375023001 Chr.15: 42723442-42723442 on Build GRCh38
rs969589428 Chr.15: 42723497-42723497 on Build GRCh38
rs540184257 Chr.15: 42723483-42723483 on Build GRCh38
rs1023892151 Chr.15: 42723482-42723482 on Build GRCh38
rs768959455 Chr.15: 42723371-42723374 on Build GRCh38
rs774242839 Chr.15: 42723648-42723648 on Build GRCh38
rs971896909 Chr.15: 42723532-42723532 on Build GRCh38
rs925448098 Chr.15: 42723534-42723534 on Build GRCh38
rs763713353 Chr.15: 42723635-42723635 on Build GRCh38
rs751250885 Chr.15: 42723689-42723690 on Build GRCh38
rs79535735 Chr.15: 42723502-42723502 on Build GRCh38
rs886922339 Chr.15: 42723644-42723644 on Build GRCh38
rs886947205 Chr.15: 42723419-42723419 on Build GRCh38
rs535251424 Chr.15: 42723462-42723462 on Build GRCh38
rs912540029 Chr.15: 42723292-42723292 on Build GRCh38
rs894062311 Chr.15: 42723458-42723458 on Build GRCh38
rs981914593 Chr.15: 42723340-42723342 on Build GRCh38
rs763302535 Chr.15: 42723627-42723627 on Build GRCh38
rs545515138 Chr.15: 42723619-42723619 on Build GRCh38
rs80066191 Chr.15: 42723668-42723668 on Build GRCh38
rs528756487 Chr.15: 42723611-42723611 on Build GRCh38
rs944066595 Chr.15: 42723309-42723309 on Build GRCh38
rs1035455818 Chr.15: 42723430-42723430 on Build GRCh38
rs969184997 Chr.15: 42723338-42723338 on Build GRCh38
rs770269083 Chr.15: 42723544-42723544 on Build GRCh38
rs1048186518 Chr.15: 42723387-42723387 on Build GRCh38
rs573659922 Chr.15: 42723664-42723664 on Build GRCh38
rs995743431 Chr.15: 42723386-42723386 on Build GRCh38
rs537036940 Chr.15: 42723564-42723564 on Build GRCh38
rs895608597 Chr.15: 42723676-42723676 on Build GRCh38

View Full Product Details