Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
FNIP2
Entrez Gene ID
57600
Gene Name
folliculin interacting protein 2
Gene Aliases
FNIPL, MAPO1
Chromosome Location
Chr.4: 158768995 - 158908050 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001323916.1 NP_001310845.1 14 - 15 3171 64
NM_020840.2 NP_065891.1 14 - 15 3166 64
XM_005263156.1 XP_005263213.1 15 - 16 3256 64
XM_005263158.1 XP_005263215.1 15 - 16 3056 64
XM_011532154.2 XP_011530456.1 15 - 16 4036 64
XM_017008487.1 XP_016863976.1 16 - 17 3205 64
GenBank mRNA AB040883.1 - 14 - 15 3023 64
AK022968.1 - 2 - 3 963 64

View Full Product Details