Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
GAPVD1
Entrez Gene ID
26130
Gene Name
GTPase activating protein and VPS9 domains 1
Gene Aliases
GAPEX5, GAPex-5, RAP6
Chromosome Location
Chr.9: 125261794 - 125365011 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001282679.1 NP_001269608.1 23 - 24 3984 74
NM_001282680.1 NP_001269609.1 23 - 24 3930 74
NM_001282681.1 NP_001269610.1 20 - 21 3535 74
NM_015635.3 NP_056450.2 22 - 23 3845 74
XM_005251901.3 XP_005251958.1 22 - 23 3794 74
XM_005251904.3 XP_005251961.1 21 - 22 3710 74
XM_006717044.3 XP_006717107.1 22 - 23 3716 74
XM_011518499.2 XP_011516801.1 23 - 24 3889 74
XM_011518500.2 XP_011516802.1 24 - 25 4006 74
XM_011518502.2 XP_011516804.1 23 - 24 3817 74
XM_011518506.2 XP_011516808.1 23 - 24 3814 74
XM_011518507.2 XP_011516809.1 22 - 23 3813 74
XM_017014600.1 XP_016870089.1 22 - 23 3699 74
XM_017014601.1 XP_016870090.1 21 - 22 3721 74
XM_017014602.1 XP_016870091.1 23 - 24 3941 74
XM_017014603.1 XP_016870092.1 23 - 24 3888 74
XM_017014604.1 XP_016870093.1 24 - 25 3990 74
XM_017014605.1 XP_016870094.1 21 - 22 3721 74
XM_017014606.1 XP_016870095.1 23 - 24 3835 74
XM_017014607.1 XP_016870096.1 23 - 24 3844 74
XM_017014608.1 XP_016870097.1 21 - 22 3654 74
XM_017014609.1 XP_016870098.1 22 - 23 3822 74
XM_017014610.1 XP_016870099.1 20 - 21 3640 74
XM_017014611.1 XP_016870100.1 21 - 22 3633 74
GenBank mRNA AB040954.3 - 23 - 24 3910 74
AK023841.1 - 14 - 15 1879 74
AK096224.1 - 4 - 5 1449 74
AL080196.1 - 12 - 13 1627 74
AL834325.1 - 2 - 3 175 74
BC013635.1 - 6 - 7 835 74
BC114937.1 - 23 - 24 3813 74
BC114962.1 - 20 - 21 3568 74
DQ233254.1 - 20 - 21 3503 74

View Full Product Details