Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.15: 43600415-43600907 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
STRC
Entrez Gene ID
161497
Gene Name
stereocilin
Gene Aliases
DFNB16
UniGene
Hs.657395
Gene Chromosome Location
Chr.15: 43599243-43619669 on Build GRCh38
Transcripts
NM_153700.2 XM_011521278.2 XM_017021954.1 XM_017021955.1 XM_017021956.1 XM_017021957.1 XM_017021958.1 XM_017021959.1 XM_017021962.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6484557 c.4968C>A p.I1656I Chr.15: 43600559-43600559 on Build GRCh38
3816155 c.4857C>G p.L1619L Chr.15: 43600670-43600670 on Build GRCh38
6457556 c.4886C>G p.S1629C Chr.15: 43600641-43600641 on Build GRCh38
1373058 c.4903G>T p.V1635F Chr.15: 43600624-43600624 on Build GRCh38
3501496 c.4809A>C p.P1603P Chr.15: 43600907-43600907 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs1010941642 Chr.15: 43600812-43600812 on Build GRCh38
rs537190397 Chr.15: 43600448-43600448 on Build GRCh38
rs775312704 Chr.15: 43600716-43600716 on Build GRCh38
rs375284306 Chr.15: 43600639-43600639 on Build GRCh38
rs2920791 Chr.15: 43600609-43600609 on Build GRCh38
rs551979899 Chr.15: 43600439-43600439 on Build GRCh38
rs12438025 Chr.15: 43600649-43600649 on Build GRCh38
rs781449029 Chr.15: 43600840-43600840 on Build GRCh38
rs1052725790 Chr.15: 43600790-43600790 on Build GRCh38
rs560371288 Chr.15: 43600797-43600797 on Build GRCh38
rs556988243 Chr.15: 43600579-43600579 on Build GRCh38
rs768401879 Chr.15: 43600876-43600876 on Build GRCh38
rs775829853 Chr.15: 43600516-43600516 on Build GRCh38
rs374759204 Chr.15: 43600733-43600733 on Build GRCh38
rs893794893 Chr.15: 43600803-43600803 on Build GRCh38
rs727503441 Chr.15: 43600609-43600610 on Build GRCh38
rs772670595 Chr.15: 43600881-43600881 on Build GRCh38
rs746259204 Chr.15: 43600515-43600515 on Build GRCh38
rs769443188 Chr.15: 43600879-43600879 on Build GRCh38
rs763598724 Chr.15: 43600590-43600590 on Build GRCh38
rs770037300 Chr.15: 43600704-43600704 on Build GRCh38
rs867795318 Chr.15: 43600778-43600778 on Build GRCh38
rs772061973 Chr.15: 43600640-43600640 on Build GRCh38
rs574386839 Chr.15: 43600793-43600793 on Build GRCh38
rs1023759444 Chr.15: 43600584-43600584 on Build GRCh38
rs2447209 Chr.15: 43600844-43600844 on Build GRCh38
rs139683375 Chr.15: 43600664-43600664 on Build GRCh38
rs775159378 Chr.15: 43600641-43600641 on Build GRCh38
rs376948482 Chr.15: 43600712-43600712 on Build GRCh38
rs961928299 Chr.15: 43600632-43600632 on Build GRCh38
rs535616758 Chr.15: 43600450-43600450 on Build GRCh38
rs370742823 Chr.15: 43600451-43600451 on Build GRCh38
rs776304785 Chr.15: 43600724-43600724 on Build GRCh38
rs12437957 Chr.15: 43600695-43600695 on Build GRCh38
rs758779420 Chr.15: 43600773-43600773 on Build GRCh38
rs1031389370 Chr.15: 43600544-43600544 on Build GRCh38
rs550560949 Chr.15: 43600599-43600599 on Build GRCh38
rs770943687 Chr.15: 43600628-43600628 on Build GRCh38
rs779178473 Chr.15: 43600509-43600509 on Build GRCh38
rs765702125 Chr.15: 43600661-43600661 on Build GRCh38
rs745860856 Chr.15: 43600707-43600707 on Build GRCh38
rs567655009 Chr.15: 43600600-43600600 on Build GRCh38
rs927800686 Chr.15: 43600617-43600617 on Build GRCh38
rs760304841 Chr.15: 43600645-43600645 on Build GRCh38
rs999717999 Chr.15: 43600786-43600786 on Build GRCh38
rs2860666 Chr.15: 43600610-43600610 on Build GRCh38
rs771993649 Chr.15: 43600864-43600864 on Build GRCh38
rs751876108 Chr.15: 43600670-43600670 on Build GRCh38
rs750994204 Chr.15: 43600663-43600663 on Build GRCh38
rs727503440 Chr.15: 43600608-43600608 on Build GRCh38
rs755576635 Chr.15: 43600580-43600580 on Build GRCh38
rs571393973 Chr.15: 43600442-43600442 on Build GRCh38
rs753288756 Chr.15: 43600594-43600594 on Build GRCh38
rs776251331 Chr.15: 43600877-43600877 on Build GRCh38
rs369759232 Chr.15: 43600526-43600526 on Build GRCh38
rs763385671 Chr.15: 43600830-43600830 on Build GRCh38
rs200327107 Chr.15: 43600568-43600568 on Build GRCh38
rs369417288 Chr.15: 43600685-43600685 on Build GRCh38
rs776707430 Chr.15: 43600622-43600622 on Build GRCh38
rs761361123 Chr.15: 43600659-43600659 on Build GRCh38
rs768328048 Chr.15: 43600722-43600722 on Build GRCh38
rs767982545 Chr.15: 43600568-43600569 on Build GRCh38
rs758801779 Chr.15: 43600606-43600606 on Build GRCh38
rs930367421 Chr.15: 43600694-43600694 on Build GRCh38
rs756252558 Chr.15: 43600845-43600845 on Build GRCh38
rs749862323 Chr.15: 43600491-43600491 on Build GRCh38
rs764784468 Chr.15: 43600734-43600734 on Build GRCh38
rs3097773 Chr.15: 43600874-43600874 on Build GRCh38
rs760405894 Chr.15: 43600717-43600717 on Build GRCh38
rs755702243 Chr.15: 43600484-43600484 on Build GRCh38
rs184909019 Chr.15: 43600682-43600682 on Build GRCh38
rs2447206 Chr.15: 43600810-43600810 on Build GRCh38
rs774483463 Chr.15: 43600633-43600633 on Build GRCh38
rs762892344 Chr.15: 43600547-43600547 on Build GRCh38
rs768845250 Chr.15: 43600536-43600536 on Build GRCh38
rs774248350 Chr.15: 43600562-43600562 on Build GRCh38
rs749407115 Chr.15: 43600857-43600857 on Build GRCh38
rs778993261 Chr.15: 43600705-43600705 on Build GRCh38
rs753336088 Chr.15: 43600655-43600655 on Build GRCh38
rs755347155 Chr.15: 43600838-43600838 on Build GRCh38
rs540126867 Chr.15: 43600795-43600795 on Build GRCh38
rs2915791 Chr.15: 43600624-43600624 on Build GRCh38
rs773011825 Chr.15: 43600546-43600546 on Build GRCh38
rs766425716 Chr.15: 43600554-43600554 on Build GRCh38
rs771905551 Chr.15: 43600711-43600711 on Build GRCh38
rs778113687 Chr.15: 43600490-43600490 on Build GRCh38
rs367587412 Chr.15: 43600741-43600741 on Build GRCh38
rs189488577 Chr.15: 43600798-43600798 on Build GRCh38
rs767260013 Chr.15: 43600571-43600571 on Build GRCh38
rs751903876 Chr.15: 43600831-43600831 on Build GRCh38
rs779901353 Chr.15: 43600866-43600866 on Build GRCh38
rs753086169 Chr.15: 43600835-43600836 on Build GRCh38
rs1041170555 Chr.15: 43600770-43600770 on Build GRCh38
rs369652730 Chr.15: 43600563-43600563 on Build GRCh38
rs757772097 Chr.15: 43600502-43600502 on Build GRCh38
rs768775965 Chr.15: 43600615-43600615 on Build GRCh38
rs2447207 Chr.15: 43600821-43600821 on Build GRCh38
rs373514051 Chr.15: 43600850-43600850 on Build GRCh38

View Full Product Details