The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001317784.1 | NP_001304713.1 | 13 - 14 | 2048 | 137 |
NM_017988.5 | NP_060458.3 | 13 - 14 | 2200 | 137 | |
XM_017019618.1 | XP_016875107.1 | 14 - 15 | 2060 | 137 | |
XM_017019619.1 | XP_016875108.1 | 14 - 15 | 2212 | 137 | |
XM_017019620.1 | XP_016875109.1 | 15 - 16 | 2199 | 137 | |
GenBank mRNA | AB037781.1 | - | 11 - 12 | 1366 | 137 |
AK001597.1 | - | 3 - 4 | 360 | 137 | |
AK024274.1 | - | 9 - 10 | 1209 | 137 | |
AK027551.1 | - | 9 - 10 | 1266 | 137 | |
AK292980.1 | - | 13 - 14 | 1974 | 137 | |
BC012387.2 | - | 4 - 5 | 493 | 137 | |
BC063798.1 | - | 13 - 14 | 1813 | 137 |