Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
SCAF1
Entrez Gene ID
58506
Gene Name
SR-related CTD associated factor 1
Gene Aliases
SRA1
Chromosome Location
Chr.19: 49640483 - 49658649 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_021228.2 NP_067051.2 3 - 4 290 77
XM_005259122.4 XP_005259179.1 3 - 4 573 77
XM_011527194.2 XP_011525496.1 3 - 4 215 77
GenBank mRNA AK024444.1 - 3 - 4 290 77
BC011662.2 - 3 - 4 194 77
BC053992.1 - 2 - 3 177 77

View Full Product Details