Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
UNC13D
Entrez Gene ID
201294
Gene Name
unc-13 homolog D
Gene Aliases
FHL3, HLH3, HPLH3, Munc13-4
Chromosome Location
Chr.17: 75827225 - 75844717 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_199242.2 NP_954712.1 22 - 23 2473 78
XM_011524504.1 XP_011522806.1 22 - 23 2106 78
XM_011524506.1 XP_011522808.1 22 - 23 2103 78
XM_011524507.2 XP_011522809.1 22 - 23 2503 78
GenBank mRNA AJ578444.1 - 22 - 23 2151 78
AK024474.1 - 20 - 21 2375 78
AK300316.1 - 23 - 24 2164 78
AK301529.1 - 22 - 23 2106 78
BC024740.2 - 20 - 21 1902 78
BC067084.1 - 22 - 23 2473 78

View Full Product Details