The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001004360.3 | NP_001004360.2 | 25 - 26 | 2364 | 102 |
NM_001178034.1 | NP_001171505.1 | 25 - 26 | 2291 | 102 | |
NM_001178036.1 | NP_001171507.1 | 26 - 27 | 2768 | 102 | |
NM_001178037.1 | NP_001171508.1 | 25 - 26 | 2334 | 102 | |
NM_001321905.1 | NP_001308834.1 | 26 - 27 | 2794 | 102 | |
NM_001321906.1 | NP_001308835.1 | 26 - 27 | 2410 | 102 | |
NM_001321907.1 | NP_001308836.1 | 24 - 25 | 2640 | 102 | |
NM_001321908.1 | NP_001308837.1 | 25 - 26 | 2653 | 102 | |
NM_001321909.1 | NP_001308838.1 | 24 - 25 | 2268 | 102 | |
NM_001321910.1 | NP_001308839.1 | 26 - 27 | 2737 | 102 | |
NM_001321911.1 | NP_001308840.1 | 25 - 26 | 2736 | 102 | |
NM_001321913.1 | NP_001308842.1 | 24 - 25 | 2564 | 102 | |
NM_001321914.1 | NP_001308843.1 | 25 - 26 | 2289 | 102 | |
NM_020868.4 | NP_065919.2 | 25 - 26 | 2679 | 102 | |
XM_017004565.1 | XP_016860054.1 | 25 - 26 | 2799 | 102 | |
XM_017004566.1 | XP_016860055.1 | 24 - 25 | 2283 | 102 | |
GenBank mRNA | AB040925.2 | - | 25 - 26 | 2246 | 102 |
AK025075.1 | - | 13 - 14 | 1090 | 102 | |
AK289967.1 | - | 25 - 26 | 2378 | 102 | |
AK296490.1 | - | 25 - 26 | 2366 | 102 | |
AY172661.3 | - | 25 - 26 | 2258 | 102 | |
AY387785.1 | - | 25 - 26 | 2280 | 102 | |
BC030832.1 | - | 25 - 26 | 2715 | 102 | |
DQ857321.1 | - | 25 - 26 | 2246 | 102 | |
DQ857322.1 | - | 25 - 26 | 2270 | 102 | |
DQ857323.1 | - | 25 - 26 | 2237 | 102 |