The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_020846.1 | NP_065897.1 | 1 - 2 | 89 | |
XM_005248121.2 | XP_005248178.1 | 1 - 2 | 89 | ||
GenBank mRNA | AB040891.1 | - | 1 - 2 | 89 | |
AK026476.1 | - | 1 - 2 | 79 | 89 | |
AK291006.1 | - | 1 - 2 | 89 | ||
BC040993.1 | - | 1 - 2 | 89 |